Learn More
BINDER™ Right Access Port (50 to 125 mm)
For Use With: BINDER units series BD
Brand: BINDER™ 8012-0050
7066.06 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- For use on right
- Includes silicone plug
- For use with a wide range of BINDER™ incubators and chambers
Specifications
Access port with silicone plug - 50 mm, right | |
Steel | |
Silicone plug |
BINDER Chambers Series BD | |
Access Port | |
50 mm |