Learn More
KleenGuard™ A45 Breathable Liquid and Particle Protection Hooded Coveralls
Single-use, breathable hooded coveralls with storm flap for respirator. The ultimate for user comfort, made from high-performance fabric and film laminate. Anti-static and general protection against water-based or chemical splashes, liquids or dust particles. PPE Category III.
Specifications
Product Type | General Purpose Coveralls |
---|---|
Gender | Unisex |
Material | KleenGuard™ |
Color | White |
Disposable | Single-use |
Description
- Supplied in packs of 25
- White, highly breathable, anti-static, silicone-free coveralls
- High-performance fabric and film laminate construction for comfort, safety and strength.
- Provides PPE Category III protection
- Storm flap and hood designed for easier taping to a respirator
- Provides anti-static and general protection against water-based or chemical splashes, liquids, and dust particles
- The ultimate combination of protection and user comfort in more critical areas.
- Hood specially designed to fit a respirator, longer arms,
- Full-length zip
Specifications
General Purpose Coveralls | |
KleenGuard™ | |
Single-use | |
Front Zipper | |
Water, Chemicals, Liquid, Particle | |
60 cm |
Unisex | |
White | |
Hooded | |
Non-sterile | |
23.4 in. | |
25/Pk. |