Learn More
Fisherbrand™ 96-Well Semi-Skirted PCR Plates
Directly compatible with all major thermal cyclers, including ABI instruments—can be used in ABI 96-well sequencers without adapters
Brand:  Fisherbrand™ 14230246
 View more versions of this product
Product Code. 11889261
  • 1000.00 NOK / Pack of 25
Only null left
In Stock
Description
Directly compatible with all major thermal cyclers, including ABI instruments—can be used in ABI 96-well sequencers without adapters
  • Virgin polypropylene
  • Clear well bottom for sample visibility
  • Raised-rim design
  • Sealable with adhesive films and foils, heat seals, and cap strips
  • 300μL maximum volume when used with adhesive and heat seals
  • Cut-away corner and alphanumeric grid
  • Cleanroom produced
Specifications
Specifications
Standard
Blue
Semi
300 μL
PCR, RT-PCR applications
Clear
8 x 12
PCR Microplate
Polypropylene
96