Learn More
Rogo Sampaic™ Stainless Steel 8-Piece Dissecting Set
A convenient and comprehensive set of high-quality dissection instruments provided in a handsome black, red-lined zippered case. Rogo Sampaic™ Stainless Steel 8-Piece Dissecting Sets are excellent for disection, surgical and general laboratory needs.
Brand:  Rogo Sampaic™ FMQ011
Product Code. 11728551
  • 109.71€ / Each
Currently Unavailable: We have no information when this product will be available.
View alternative products
Description
Description
  • 1 pair of strong, sharp/round 140mm scissors
  • 1 pair of Iris 110mm scissors
  • 1 pair of strong rounded 140mm pliers
  • 1 pair of find rounded 110mm tweezers/forceps
  • 1 No. 4 scalpel handle
  • 1 sachet of 5 scalpel blades, No. 23
  • 2 straight-pointed needles for dissociating stitches
Specifications
Specifications
1 Ea.
Dissecting Kit, 8 Piece