Learn More
7-Chloroquinoline, 97%, Thermo Scientific™
Product Code. p-7077595 Shop All Thermo Scientific Chemicals Products
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
5 g
missing translation for 'unitSize'
1g
5g
Chemical Identifiers
CAS 612-61-3
Molecular Formula C9H6ClN
Molecular Weight (g/mol) 163.61
InChI Key QNGUPQRODVPRDC-UHFFFAOYSA-N
Synonym quinoline, 7-chloro, 7-chloro-quinoline, pubchem19899
PubChem CID 521963
IUPAC Name 7-chloroquinoline
SMILES C1=CC2=C(C=C(C=C2)Cl)N=C1
Specifications
Assay Percent Range 97%
Packaging Glass Bottle
Quantity 1 g
Formula Weight 163.61
Percent Purity 97%
Chemical Name or Material 7-Chloroquinoline
compliance-icons
Product Identifier
  • 7-Chloroquinoline, 97%
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.