Learn More
Molecular Probes™ 6-TAMRA, SE (6-Carboxytetramethylrhodamine, Succinimidyl Ester), single isomer
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
Description
The isomeric 6-TAMRA, SE is an amine-reactive form of tetramethylrhodamine. The single-isomer preparations of TAMRA are most important for high-resolution techniques such as DNA sequencing and capillary electrophoresis. 6-TAMRA is one of the traditional fluorophores used for automated DNA sequencing.
Specifications
Specifications
Quantity | 5 mg |
Product Type | 6-TAMRA SE |
Content And Storage | Store in freezer (-5 to -30°C) and protect from light. |
Chemical Reactivity | Amine |
Shipping Condition | Room Temperature |
Reactive Moiety | Active Ester, Succinimidyl Ester |
Label or Dye | TAMRA™ Isomers, TMR (Tetramethylrhodamine) |
Label Type | Classic Dyes |