Learn More
Greiner Bio-One 50 mL Sterile Serological Pipets
Drip-free pipetting design with high optical clarity and maximum accuracy.
144.29 €
Specifications
Material | Polystyrene |
---|---|
Sterility | Sterile |
Graduated | Yes |
Graduations | 1/2 |
Volume (Metric) | 50 mL |
Description
- High-grade polystyrene
- Clear, easily legible graduation
- Colored vertical stripe for ease in reading the volume
- With synthetic fibrous filter material for protection against
Specifications
Polystyrene | |
Yes | |
50 mL |
Sterile | |
1/2 | |
Serological Pipet |