Learn More
5-Norbornen-2-ol, mixture of endo and exo, 98+%, Thermo Scientific Chemicals
Product Code. 15473548
Click to view available options
Quantity:
1 g
5 g
25 g
It is applied in High Performance Liquid Chromatography. (HPLC) and Nuclear Magnetic Resonance (NMR).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is applied in High Performance Liquid Chromatography. (HPLC) and Nuclear Magnetic Resonance (NMR).
Solubility
Slightly soluble in water
Notes
Store at 2-8°C. Incompatible with oxidizing agents and heat.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 13080-90-5
Molecular Formula C7H10O
Molecular Weight (g/mol) 110.156
MDL Number MFCD00167566
InChI Key MKOSBHNWXFSHSW-UHFFFAOYSA-N
Synonym 5-norbornene-2-ol, bicyclo 2.2.1 hept-5-en-2-ol, 5-norbornen-2-ol, 5-norbornen-2-ol, exo, exo-2-norbornenol, bicyclo 2.2.1 hept-2-en-5-ol, bicyclo 2.2.1 hept-5-en-2-ol, exo, 5-norbornen-2-ol exo
PubChem CID 96066
IUPAC Name bicyclo[2.2.1]hept-2-en-5-ol
SMILES C1C2CC(C1C=C2)O
Specifications
Melting Point 104°C to 106°C
Flash Point 51°C (124°F)
Quantity 5 g
UN Number UN1325
Solubility Information Slightly soluble in water
Formula Weight 110.15
Percent Purity 99%
Chemical Name or Material 5-Norbornen-2-ol, mixture of endo and exo