Learn More
5-Iodoisatin, 97%, Thermo Scientific Chemicals
Product Code. 11447207
Click to view available options
Quantity:
5 g
25 g
100 g
5-Iodoisatin undergoes condensation reaction with phenol yields 5-iodophenolisatin, malonic acid yields 6-iodo-2-quinolone-4-carboxylic acid.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
5-Iodoisatin undergoes condensation reaction with phenol yields 5-iodophenolisatin, malonic acid yields 6-iodo-2-quinolone-4-carboxylic acid.
Solubility
Soluble acetone, acetic acid, ethanol. Insoluble in water.
Notes
Light Sensitive. Keep container tightly closed. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 20780-76-1
Molecular Formula C8H4INO2
Molecular Weight (g/mol) 273.029
MDL Number MFCD00016899
InChI Key OEUGDMOJQQLVAZ-UHFFFAOYSA-N
Synonym 5-iodoisatin, 5-iodoindoline-2,3-dione, 5-iodo isatin, 5-iodoindole-2,3-dione, 5-iodo-2,3-dioxoindoline, 1h-indole-2,3-dione, 5-iodo, 5-iodo-2,3-dihydro-1h-indole-2,3-dione, 5-iodo-1h-benzo d azolidine-2,3-dione, 5-iodo-isatine, 5-iodo-isatin
PubChem CID 88695
IUPAC Name 5-iodo-1H-indole-2,3-dione
SMILES C1=CC2=C(C=C1I)C(=O)C(=O)N2
Specifications
Melting Point ∼276°C (decomposition)
Quantity 25 g
Beilstein 131248
Sensitivity Light sensitive
Solubility Information Soluble acetone,acetic acid,ethanol. Insoluble in water.
Formula Weight 273.03
Percent Purity 97%
Chemical Name or Material 5-Iodoisatin