Learn More
5-(Ethoxycarbonyl)pentylzinc bromide, 0.5M in THF, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific™
Product Code. 15455345
Click to view available options
Quantity:
50 mL
Unit Size:
50mL
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Specifications
CAS 312693-01-9
Density 0.98
Molecular Formula C8H15BrO2Zn
Linear Formula CH3CH2O2C(CH2)5ZnBr
MDL Number MFCD00671979
Quantity 50 mL
UN Number UN3399
Sensitivity Air sensitive; May form a precipitate
Synonym zinc 2+ ion 6-ethoxy-6-oxohexan-1-ide bromide
InChI Key ZISDBYYELZLQGH-UHFFFAOYSA-M
SMILES CCOC(=O)CCCC[CH2-].[Zn+2].[Br-]
IUPAC Name zinc;ethyl hexanoate;bromide
Molecular Weight (g/mol) 288.49
PubChem CID 57377104
Formula Weight 288.49
Chemical Name or Material 5-(Ethoxycarbonyl)pentylzinc bromide, Packaged under Argon in resealable ChemSeal™ bottles