Learn More
3M™ 4432+ Lab Coats
Designed to help protect the wearer against light splashing with non-hazardous liquids and dust. 3M™ 4432+ Lab Coats are made from super-breathable SSMMS non-woven for effective release of body heat and moisture.
2146.16 NOK
Specifications
Product Type | General Purpose Lab Coat |
---|---|
Gender | Unisex |
Material | Polypropylene, Polyethylene Laminate |
Color | Cool White |
Cuff Style | Knit |
Description
- Anti-static
- Front zip closing
- Low lint properties
- Reduced heat stress
- Knitted cuffs and collared
- Internal pocket on the wearer's left-hand side
Specifications
General Purpose Lab Coat | |
Polypropylene, Polyethylene Laminate | |
Knit | |
Non-sterile | |
CE Category 1 |
Unisex | |
Cool White | |
Reusable | |
Serged | |
Single Pack |