Learn More
Labconco 4 place Microplate Rotor - CentriVap Pro
Rotors for use with Labconco™ CentriVap™ Pro Vacuum Concentrators. Stainless steel and PTFE coated options available.
9301.52 DKK - 16044.26 DKK
Specifications
Material | Stainless Steel |
---|---|
Type | Rotor |
Rotor System | CentriVap Pro |
Dimensions (L x W x H) | 25.4 x 15.75 x 7.62 cm (10 x 6.2 x 3 in.) |
Product Code | Brand | Material (Coating) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Material (Coating) | Price | Quantity & Availability | |||||
30533406
![]() |
Labconco
7555800 |
9301.52 DKK
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
30533405
![]() |
Labconco
7555801 |
PTFE |
16044.26 DKK
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- Compatible with CentriVap Pro Vacuum Concentrators
- Stainless steel material
- Optional PTFE coated
Specifications
Stainless Steel | |
Rotor | |
CentriVap Pro | |
25.4 x 15.75 x 7.62 cm (10 x 6.2 x 3 in.) |