Learn More
4-Fluorobenzoyl chloride, 98%
Product Code. p-7020852
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
CAS: 403-43-0 | C7H4ClFO | 158.556 g/mol
It is used in the synthesis of polyphenylene ether and thioether ketones and also in the synthesis fluorinated poly(aryl ether ketone)s containing 1,4-naphthalene moieties. It is also used as a raw material in engineering of plastics.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is used in the synthesis of polyphenylene ether and thioether ketones and also in the synthesis fluorinated poly(aryl ether ketone)s containing 1,4-naphthalene moieties. It is also used as a raw material in engineering of plastics.
Solubility
Reacts with water.
Notes
Moisture sensitive. Store away from bases, water/moisture and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under gas.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 403-43-0
Molecular Formula C7H4ClFO
Molecular Weight (g/mol) 158.556
MDL Number MFCD00000684
InChI Key CZKLEJHVLCMVQR-UHFFFAOYSA-N
Synonym benzoyl chloride, 4-fluoro, p-fluorobenzoyl chloride, benzoyl chloride, p-fluoro, p-fluorobenzoic acid chloride, 4-fluorobenzoylchloride, fluorobenzoyl 4-chloride, 4-fluorobenzoic acid chloride, p-fluorobenzoylchloride, p-fluorobenzoyl choride, 4-flurobenzoyl chloride
PubChem CID 67879
IUPAC Name 4-fluorobenzoyl chloride
SMILES C1=CC(=CC=C1C(=O)Cl)F
Specifications
Melting Point 10°C to 12°C
Density 1.326
Boiling Point 193°C
Flash Point 82°C (179°F)
Odor Acidic
Refractive Index 1.532
Quantity 25 g
UN Number UN3265
Beilstein 386215
Sensitivity Moisture sensitive
Solubility Information Reacts with water.
Formula Weight 158.56
Percent Purity 98%
Chemical Name or Material 4-Fluorobenzoyl chloride