Learn More
4-Chloro-O-tolylhydrazine hydrochloride, 97%, Thermo Scientific™
713.00 NOK - 3375.00 NOK
Chemical Identifiers
CAS | 19690-59-6 |
---|---|
Molecular Formula | C7H10Cl2N2 |
Molecular Weight (g/mol) | 193.071 |
MDL Number | MFCD00012944 |
InChI Key | VBVZXWFQHFXJJK-UHFFFAOYSA-N |
Synonym | 4-chloro-2-methylphenyl hydrazine hydrochloride, 1-4-chloro-2-methylphenyl hydrazine hydrochloride, 4-chloro-2-tolylhydrazine hydrochloride, 4-chloro-2-methylphenylhydrazine, chloride, 4-chloro-2-tolylhydrazine hcl, 5-chloro-2-hydrazinotoluene hydrochloride, hydrazine, 4-chloro-2-methylphenyl-, hydrochloride 1:1 |
PubChem CID | 3084321 |
IUPAC Name | (4-chloro-2-methylphenyl)hydrazine;hydrochloride |
SMILES | CC1=C(C=CC(=C1)Cl)NN.Cl |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10169222
|
Thermo Scientific Maybridge
SB01504DA |
1 g |
713.00 NOK
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10022422
|
Thermo Scientific Maybridge
SB01504EA |
10 g |
1880.00 NOK
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10072404
|
Thermo Scientific Maybridge
SB01504EB |
25 g |
3375.00 NOK
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Chemical Identifiers
19690-59-6 | |
193.071 | |
VBVZXWFQHFXJJK-UHFFFAOYSA-N | |
3084321 | |
CC1=C(C=CC(=C1)Cl)NN.Cl |
C7H10Cl2N2 | |
MFCD00012944 | |
4-chloro-2-methylphenyl hydrazine hydrochloride, 1-4-chloro-2-methylphenyl hydrazine hydrochloride, 4-chloro-2-tolylhydrazine hydrochloride, 4-chloro-2-methylphenylhydrazine, chloride, 4-chloro-2-tolylhydrazine hcl, 5-chloro-2-hydrazinotoluene hydrochloride, hydrazine, 4-chloro-2-methylphenyl-, hydrochloride 1:1 | |
(4-chloro-2-methylphenyl)hydrazine;hydrochloride |
Specifications
19690-59-6 | |
97% | |
C7H10Cl2N2 | |
MFCD00012944 | |
VBVZXWFQHFXJJK-UHFFFAOYSA-N | |
(4-chloro-2-methylphenyl)hydrazine;hydrochloride | |
3084321 | |
97% |
97.0 | |
Amber glass bottle | |
ClC6H3(CH3)NHNH2·HCl | |
4-chloro-2-methylphenyl hydrazine hydrochloride, 1-4-chloro-2-methylphenyl hydrazine hydrochloride, 4-chloro-2-tolylhydrazine hydrochloride, 4-chloro-2-methylphenylhydrazine, chloride, 4-chloro-2-tolylhydrazine hcl, 5-chloro-2-hydrazinotoluene hydrochloride, hydrazine, 4-chloro-2-methylphenyl-, hydrochloride 1:1 | |
CC1=C(C=CC(=C1)Cl)NN.Cl | |
193.071 | |
193.08 | |
1-(4-Chloro-2-methylphenyl)hydrazine hydrochloride |
Safety and Handling
GHS H Statement:
Harmful in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning