Learn More
4-(Aminomethyl)indole, 97%
Product Code. p-7036643
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
CAS: 3468-18-6 | C9H10N2 | 146.19 g/mol
4-(Aminomethyl)indole is used as a reactant for preparation of dopamine receptor antagonists and for preparation of high-affinity ligands to α2δ subunit of voltage-gated calcium channels. It is used as a synthetic intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-(Aminomethyl)indole is used as a reactant for preparation of dopamine receptor antagonists and for preparation of high-affinity ligands to α2δ subunit of voltage-gated calcium channels. It is used as a synthetic intermediate.
Solubility
Soluble in ethanol, dimethyl sulfoxide and dimethyl formamide.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 3468-18-6
Molecular Formula C9H10N2
Molecular Weight (g/mol) 146.19
MDL Number MFCD04973297
InChI Key FFBWKPKOXRMLNP-UHFFFAOYSA-N
Synonym 4-aminomethylindole, 1h-indol-4-yl methanamine, 1h-indole-4-methanamine, 4-aminomethyl indole, 1h-indol-4-ylmethyl amine, 1-1h-indol-4-yl methanamine, c-1h-indol-4-yl-methylamine, indole-4-methylamine, 4-aminomethyl-indole, 4-aminomethyl-1h-indole
PubChem CID 280302
IUPAC Name 1H-indol-4-ylmethanamine
SMILES NCC1=C2C=CNC2=CC=C1
Specifications
Melting Point 130°C to 135°C
Quantity 250 mg
Solubility Information Soluble in ethanol,dimethyl sulfoxide and dimethyl formamide.
Formula Weight 146.19
Percent Purity 97%
Chemical Name or Material 4-(Aminomethyl)indole