Learn More
4-(1-Bromoethyl)benzoic acid, 98%
Product Code. p-7046072
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
CAS: 113023-73-7 | C9H9BrO2 | 229.073 g/mol
4-(1-Bromoethyl)benzoic acid acts as a reagent to prepare 4-(1-bromo-ethyl)-benzoic acid ethyl ester. It is also used as an intermediate for active pharmaceutical ingredient as well as in chemical synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-(1-Bromoethyl)benzoic acid acts as a reagent to prepare 4-(1-bromo-ethyl)-benzoic acid ethyl ester. It is also used as an intermediate for active pharmaceutical ingredient as well as in chemical synthesis.
Solubility
Insoluble in water.
Notes
Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 113023-73-7
Molecular Formula C9H9BrO2
Molecular Weight (g/mol) 229.073
MDL Number MFCD00020378
InChI Key VICCYULHZWEWMB-UHFFFAOYSA-N
Synonym 4-1-bromoethyl benzoic acid, benzoic acid, 4-1-bromoethyl, acmc-1c3tf, 4-1-bromoethyl-benzoic acid, 4-1-bromo-ethyl-benzoic acid, benzoic acid,4-1-bromoethyl
PubChem CID 5146225
IUPAC Name 4-(1-bromoethyl)benzoic acid
SMILES CC(C1=CC=C(C=C1)C(=O)O)Br
Specifications
Melting Point 155°C to 156°C
Quantity 1 g
UN Number UN3261
Beilstein 2084039
Solubility Information Insoluble in water.
Formula Weight 229.08
Percent Purity 98%
Chemical Name or Material 4-(1-Bromoethyl)benzoic acid