Learn More
3-Iodo-1-methyl-1H-pyrazole, 97%, Thermo Scientific Chemicals
Product Code. 15482729
Click to view available options
Quantity:
250 mg
1 g
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 92525-10-5
Molecular Formula C4H5IN2
Molecular Weight (g/mol) 208.002
MDL Number MFCD08435890
InChI Key SITJXRWLFQGWCB-UHFFFAOYSA-N
Synonym 3-iodo-1-methyl-1h-pyrazole, 1-methyl-3-iodopyrazole, 1h-pyrazole, 3-iodo-1-methyl, 3-iodo-1-methy-1h-pyrazole, iodo-n-methylpyrazole, 3-idio-1-methylpyrazole, 3-iodanyl-1-methyl-pyrazole, 1h-pyrazole,3-iodo-1-methyl, 3 5-iodopyrazole, 1-me
PubChem CID 13334057
IUPAC Name 3-iodo-1-methylpyrazole
SMILES CN1C=CC(=N1)I
Specifications
Color Brown
Density 1.936 g/mL
Flash Point 110°C (230°F)
Refractive Index 1.588
Quantity 250 mg
Sensitivity Light sensitive
Formula Weight 208
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 3-Iodo-1-methyl-1H-pyrazole