Learn More
3-Chloro-2,4-difluorophenylacetic acid, 97%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
3-Chloro-2,4-difluorophenylacetic acid is used as an intermediate in organic synthesis.
Notes
Incompatible with oxidizing agents.
3-Chloro-2,4-difluorophenylacetic acid is used as an intermediate in organic synthesis.
Notes
Incompatible with oxidizing agents.
Specifications
Specifications
Melting Point | 107°C to 110°C |
Quantity | 250 mg |
Formula Weight | 206.58 |
Percent Purity | 97% |
Chemical Name or Material | 3-Chloro-2,4-difluorophenylacetic acid |