missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-1-propanol, 98%, stab.

Product Code. 11402908
500 g, 500g
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
This item is not returnable. View return policy
This item is not returnable. View return policy

CAS: 627-30-5 | C3H7ClO | 94.538 g/mol

It is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a pharmaceutical intermediate.

Solubility
Soluble in water at 20°C 300g/L.

Notes
Store away from oxidizing agents and bases. Keep the container tightly sealed and store in cool, dry condition in well ventilated place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 627-30-5
Molecular Formula C3H7ClO
Molecular Weight (g/mol) 94.538
MDL Number MFCD00002943
InChI Key LAMUXTNQCICZQX-UHFFFAOYSA-N
Synonym 3-chloro-1-propanol, 3-chloropropanol, 1-propanol, 3-chloro, 1-chloro-3-hydroxypropane, trimethylene chlorohydrin, 3-chloro-l-propanol, 3-chlorpropan-1-ol, 3-choro-1-propanol, 1-chloro-3-propanol, 3-chloropropanol-1
PubChem CID 12313
IUPAC Name 3-chloropropan-1-ol
SMILES C(CO)CCl

Specifications

Melting Point -20°C
Density 1.131
pH 3 to 4
Boiling Point 160°C to 162°C
Flash Point 73°C (163°F)
Odor Pleasant
Refractive Index 1.445
Quantity 500 g
UN Number UN2849
Beilstein 773655
Solubility Information Soluble in water at 20°C 300g/L.
Formula Weight 94.54
Percent Purity 98%
Chemical Name or Material 3-Chloro-1-propanol
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
3-Chloro-1-propanol, 98%, stab. >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.