missing translation for 'onlineSavingsMsg'
Learn More

3-Acetoxybenzonitrile, 97%, Thermo Scientific™

Product Code. 15450468
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 55682-11-6

Specifications

Molecular Formula C7H9BO2
MDL Number MFCD00039138
Quantity 1 g
Beilstein 2576295
Synonym 4-tolylboronic acid, p-tolylboronic acid, 4-methylphenyl boronic acid, p-tolueneboronic acid, 4-methylbenzeneboronic acid, p-methylbenzeneboronic acid, boronic acid, 4-methylphenyl, boronic acid, p-tolyl, 4-methylphenylboric acid, tolyl boronic acid
InChI Key BIWQNIMLAISTBV-UHFFFAOYSA-N
SMILES CC1=CC=C(C=C1)B(O)O
IUPAC Name (4-methylphenyl)boronic acid
Molecular Weight (g/mol) 135.96
PubChem CID 79799
Formula Weight 161.16
Percent Purity 97%
Chemical Name or Material 3-Acetoxybenzonitrile
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.