Learn More
3,3-Diethoxy-1-propylboronic acid pinacol ester, 97%, Thermo Scientific Chemicals
1090.00 NOK - 3290.00 NOK
Chemical Identifiers
CAS 165904-27-8
Molecular Formula C13H27BO4
Molecular Weight (g/mol) 258.165
MDL Number MFCD03788723
InChI Key VEDSPUPKZZMMLL-UHFFFAOYSA-N
Synonym 3,3-diethoxy-1-propylboronic acid pinacol ester, 2-3,3-diethoxypropyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 4,4,5,5-tetramethyl-1,3,2-dioxaborolane-2-propanal diethyl acetal
PubChem CID 12115795
IUPAC Name 2-(3,3-diethoxypropyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
SMILES B1(OC(C(O1)(C)C)(C)C)CCC(OCC)OCC
View More Specs
Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11388906
SDS View Documents
Thermo Scientific Alfa Aesar
L19532.MD
250 mg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11378906
SDS View Documents
Thermo Scientific Alfa Aesar
L19532.03
1 g
3290.00 NOK
1g
Description
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Chemical Identifiers
165904-27-8
258.165
VEDSPUPKZZMMLL-UHFFFAOYSA-N
12115795
B1(OC(C(O1)(C)C)(C)C)CCC(OCC)OCC
C13H27BO4
MFCD03788723
3,3-diethoxy-1-propylboronic acid pinacol ester, 2-3,3-diethoxypropyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 4,4,5,5-tetramethyl-1,3,2-dioxaborolane-2-propanal diethyl acetal
2-(3,3-diethoxypropyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Specifications
165904-27-8
1.428
3,3-diethoxy-1-propylboronic acid pinacol ester, 2-3,3-diethoxypropyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 4,4,5,5-tetramethyl-1,3,2-dioxaborolane-2-propanal diethyl acetal
B1(OC(C(O1)(C)C)(C)C)CCC(OCC)OCC
258.165
258.17
3,3-Diethoxy-1-propylboronic acid pinacol ester
C13H27BO4
MFCD03788723
VEDSPUPKZZMMLL-UHFFFAOYSA-N
2-(3,3-diethoxypropyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
12115795
97%
Videos
Safety and Handling
Safety and Handling
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures