Learn More
(3,3,3-Trifluoropropyl)trichlorosilane, 97%, Thermo Scientific Chemicals
Product Code. p-7046877
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
(3,3,3-Trifluoropropyl)trichlorosilane is used for the production of fluoro silicones liquids.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
(3,3,3-Trifluoropropyl)trichlorosilane is used for the production of fluoro silicones liquids.
Solubility
Reacts with water.
Notes
Moisture Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 592-09-6
Molecular Formula C3H4Cl3F3Si
Molecular Weight (g/mol) 231.50
MDL Number MFCD00013602
InChI Key WEUBQNJHVBMUMD-UHFFFAOYSA-N
Synonym trichloro 3,3,3-trifluoropropyl silane, 3,3,3-trifluoropropyl trichlorosilane, silane, trichloro 3,3,3-trifluoropropyl, 3,3,3-trifluoropropyltrichlorosilane, acmc-20ajez, cf3ch2ch2sicl3, silane, trichloro trifluoropropyl, 3,3,3-trifluropropyltrichlorosiline, trichloro 3,3,3-trifluoropropyl silane #
PubChem CID 68963
IUPAC Name trichloro(3,3,3-trifluoropropyl)silane
SMILES FC(F)(F)CC[Si](Cl)(Cl)Cl
Specifications
Boiling Point 113°C to 114°C
Quantity 5 g
Linear Formula CF3CH2CH2SiCl3
UN Number UN2985
Beilstein 1748876
Flash Point 15°C (59°F)
Solubility Information Reacts with water.
Formula Weight 231.51
Percent Purity 97%
Refractive Index 1.386
Sensitivity Moisture sensitive
Density 1.395
Chemical Name or Material (3,3,3-Trifluoropropyl)trichlorosilane