Learn More
Macherey-Nagel™ 3 in 1 Swimming Pool Test
Brand:  Macherey-Nagel™ 90752
25.30 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Product Code. 10259691
  • 25.95€ / Pack of 50
Only null left
In Stock
Specifications
6.4 to 8.4
Determination of the water quality
Free chlorine, Alkalinity, pH
30 sec.
Strip
0, 0.5, 1, 3, 5, 10mg/L Cl2 Free Chlorine, 0, 80, 120, 180, 240mg/L CaCO3 Alkalinity, 6.4, 6.8, 7.2, 7.6, 8.4 pH
50 Pack
Swimming Pool Test