Learn More
3M™ Polycarbonate Protective Safety Glasses, 2720 Series
Click to view available options
Lens Tint:
Amber
Clear
Gray
Unit Size:
Each
Pack of 20
Description
Extremely lightweight and comfortable with a sporty, modern design.
- Optical Class 1 for high optical clarity
- Polycarbonate lens provides reliable protection against specified levels of hazardous UV radiation
- Anti-scratch coating for increased durability and improved vision
- Anti-fog coating to help reduce lens fogging, particularly during physical work or when wearing half masks
- Ideal for use in many environments, including agriculture, assembly work, automotive and bodyshops, construction, DIY, grinding, laboratories, light engineering, and woodworking
- Combines well with 3M™ Aura™ 9300+ Series Disposable Respirators
Specifications
Specifications
Lens Coating | Anti-fog, Scratch-resistant |
Lens Tint | Clear |
Product Type | Safety glasses |
For Use With (Application) | Construction, Metal Fabrication, forestry |
Frame Color | Blue |
No. per Pack | 1 Ea. |
Lens Material | Polycarbonate |
Product Content Correction
Product Title
3M™ Polycarbonate Protective Safety Glasses, 2720 Series >