Learn More
2-Valerylthiophene, 97%, Thermo Scientific™
Product Code. p-7028911
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 53119-25-8
Molecular Formula C9H12OS
Molecular Weight (g/mol) 168.254
MDL Number MFCD00053149
InChI Key KJMPRCVRMAHTBR-UHFFFAOYSA-N
Synonym 1-thien-2-ylpentan-1-one, 1-2-thienyl pentan-1-one, 1-thiophen-2-yl pentan-1-one, 1-pentanone, 1-2-thienyl, unii-x71z1k8ray, 2-valerylthiophene, x71z1k8ray, 2-pentanoylthiophene, butyl 2-thienyl ketone, acmc-20aors
PubChem CID 574100
IUPAC Name 1-thiophen-2-ylpentan-1-one
SMILES CCCCC(=O)C1=CC=CS1
Specifications
Refractive Index 1.5358
Quantity 1 g
Formula Weight 168.25
Percent Purity 97%
Chemical Name or Material 2-Valerylthiophene