Learn More
2-Trimethylsilyl-1-ethylboronic acid pinacol ester, 97%, Thermo Scientific™
Product Code. 11318916
Click to view available options
Quantity:
250 mg
1 g
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 165904-20-1
Molecular Formula C11H25BO2Si
Molecular Weight (g/mol) 228.21
MDL Number MFCD03788725
InChI Key YKYAZAJUMAWTGO-UHFFFAOYSA-N
Synonym 2-trimethylsilyl-1-ethylboronic acid pinacol ester, trimethyl 2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl ethyl silane, trimethyl 2-tetramethyl-1,3,2-dioxaborolan-2-yl ethyl silane
PubChem CID 11436111
IUPAC Name trimethyl-[2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)ethyl]silane
SMILES CC1(C)OB(CC[Si](C)(C)C)OC1(C)C
Specifications
Refractive Index 1.431
Quantity 1 g
Beilstein 8683473
Sensitivity Moisture sensitive
Formula Weight 228.22
Percent Purity 97%
Chemical Name or Material 2-Trimethylsilyl-1-ethylboronic acid pinacol ester