missing translation for 'onlineSavingsMsg'
Learn More

(+/-)-2-(Trifluoromethyl)pyrrolidine, 95%

Product Code. p-7028506
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
This item is not returnable. View return policy
This item is not returnable. View return policy

CAS: 109074-67-1 | C5H8F3N | 139.121 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 109074-67-1
Molecular Formula C5H8F3N
Molecular Weight (g/mol) 139.121
MDL Number MFCD02663405
InChI Key VINAMCOZNJHNIH-UHFFFAOYSA-N
Synonym 2-trifluoromethyl pyrrolidine, 2-trifluoromethyl-pyrrolidine, dl-2-trifluoromethylpyrrolidine, pyrrolidine,2-trifluoromethyl, pyrrolidine, 2-trifluoromethyl, +/--2-trifluoromethyl pyrrolidine, acmc-20dptq, acmc-1cih7, 2-trifluoromethyl tetrahydropyrrole
PubChem CID 2782839
IUPAC Name 2-(trifluoromethyl)pyrrolidine
SMILES C1CC(NC1)C(F)(F)F

Specifications

Density 1.21
Boiling Point 110°C to 112°C
Flash Point 26°C (78°F)
Refractive Index 1.379
Quantity 250 mg
UN Number UN2733
Sensitivity Air Sensitive
Formula Weight 139.12
Percent Purity 95%
Chemical Name or Material (+/-)-2-(Trifluoromethyl)pyrrolidine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.