Learn More
2-Propanol, ≥99.5%, Honeywell Riedel-de Haën™
Product Code. p-7120077
Click to view available options
Quantity:
1 L
4 L
Packaging:
Glass Bottle
Unit Size:
1 L
4 L
ACS Reagent,≥99.5%
ACS GENERAL USE SOLVENTS
  • value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • suitable for less demanding applications such as routine isocratic HPLC
  • variations and decrease issues traceable to solvent contamination
  • all solvents are tested to meet ACS general use specifications
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 67-63-0
Molecular Formula C3H8O
Molecular Weight (g/mol) 60.096
MDL Number MFCD00011674
InChI Key KFZMGEQAYNKOFK-UHFFFAOYSA-N
Synonym isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol
PubChem CID 3776
ChEBI CHEBI:17824
IUPAC Name propan-2-ol
SMILES CC(C)O
Specifications
pH Neutral
Melting Point -90°C
Density 0.785g/mL at 25°C
Boiling Point 77°C to 83°C
Quantity 1 L
Explosion Limit 0.02
Ignition Point 750°C
Odor Alcohol-like
Packaging Glass Bottle
Physical Form Liquid
Linear Formula (CH3)2CHOH
Vapor Pressure 33mmHg (20°C), 44mmHg (25°C)
UN Number UN1219
Beilstein 635639
Formula Weight 60.1g/mol
Percent Purity ≥99.5%
Chemical Name or Material 2-Propanol