Learn More
Lenz Laborglasintrumente 2-Neck Adapters
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
325.00 NOK - 466.00 NOK
Specifications
Material Duran Glass
View More Specs
Products 3
Product Code Brand Standard Taper Joint No. Price Quantity & Availability  
Product Code Brand Standard Taper Joint No. Price Quantity & Availability  
10221411
View Documents
Lenz Laborglasintrumente
05000111
Socket Vertical NS 14/23
325.00 NOK
Each
Estimated Shipment: 06-05-2025
to see stock.
Add to basket
10244581
View Documents
Lenz Laborglasintrumente
05000222
Socket Vertical NS 19/26
466.00 NOK
Each
Estimated Shipment: 06-05-2025
to see stock.
Add to basket
10366071
View Documents
Lenz Laborglasintrumente
05000414
Socket Vertical NS 29/32
385.00 NOK
Each
Estimated Shipment: 06-05-2025
Description
  • With two NS sockets and one NS cone
  • Ascending angle: 105°
  • Made of DURAN™ tubing
Specifications
Specifications
Duran Glass