Learn More
2-Bromo-5-fluorophenol, 97% Product Code.: 11437347
2-Bromo-5-fluorophenol, 97%
Product Code. 11437347
100 g, 100g
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
Product Code. 11437347
Brand: Thermo Scientific Alfa Aesar A15064.22
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
CAS: 147460-41-1 | C6H4BrFO | 190.999 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 147460-41-1
Molecular Formula C6H4BrFO
Molecular Weight (g/mol) 190.999
MDL Number MFCD00040939
InChI Key HUVAOAVBKOVPBZ-UHFFFAOYSA-N
Synonym phenol, 2-bromo-5-fluoro, 2-bromo-5-fluoro-phenol, pubchem1964, 2-bromo-5-fluorphenol, acmc-209cyi, 2-bromo-5-fluorphenol;, ksc493i7h, paragos 390207, 2-bromo-5-fluorophenol, attercop-chm at106843
PubChem CID 2724600
IUPAC Name 2-bromo-5-fluorophenol
SMILES C1=CC(=C(C=C1F)O)Br
Specifications
Density 1.717
Boiling Point 184°C to 186°C
Flash Point 87°C (188°F)
Refractive Index 1.554
Quantity 100 g
Beilstein 7915214
Formula Weight 191
Percent Purity 97%
Chemical Name or Material 2-Bromo-5-fluorophenol
Product Title
2-Bromo-5-fluorophenol, 97% >