missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-2'-nitroacetophenone, 98%, Thermo Scientific Chemicals

Product Code. p-3790874 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6851-99-6
Molecular Formula C8H6BrNO3
Molecular Weight (g/mol) 244.4
MDL Number MFCD00010294
InChI Key SGXUUCSRVVSMGK-UHFFFAOYSA-N
Synonym 2-bromo-2'-nitroacetophenone, 2'-nitrophenacyl bromide, 2-bromo-1-2-nitrophenyl ethanone, 2-nitrophenacyl bromide, 2-bromo-1-2-nitro-phenyl-ethanone, o-nitrophenacyl bromide, o-nitrophenethyl bromide, 2-o-nitrophenyl ethyl bromide, 2-bromo-1-2-nitrophenyl ethan-1-one, ethanone, 2-bromo-1-2-nitrophenyl
PubChem CID 244025
IUPAC Name 2-bromo-1-(2-nitrophenyl)ethanone
SMILES C1=CC=C(C(=C1)C(=O)CBr)[N+](=O)[O-]

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 55.0°C to 57.0°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula O2NC6H4COCH2Br
Quantity 1 g
Formula Weight 244.4
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 2-Bromo-2'-nitroacetophenone
Show More Show Less
compliance-icons
Product Identifier
  • 2-Bromo-2'-nitroacetophenone
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.