missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-6-chlorobenzothiazole, 99%

Product Code. p-7030310
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy
This item is not returnable. View return policy

CAS: 95-24-9 | C7H5ClN2S | 184.641 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 95-24-9
Molecular Formula C7H5ClN2S
Molecular Weight (g/mol) 184.641
MDL Number MFCD00053557
InChI Key VMNXKIDUTPOHPO-UHFFFAOYSA-N
Synonym 2-amino-6-chlorobenzothiazole, 6-chlorobenzothiazol-2-ylamine, 2-benzothiazolamine, 6-chloro, 6-chlorobenzo d thiazol-2-amine, 6-chloro-benzothiazol-2-ylamine, unii-2u337t5ufg, 6-chloro-2-benzothiazolamine, benzothiazole, 2-amino-6-chloro, 6-chlorobenzothiazole-2-ylamine, c7h4brcln2s
PubChem CID 7226
IUPAC Name 6-chloro-1,3-benzothiazol-2-amine
SMILES C1=CC2=C(C=C1Cl)SC(=N2)N

Specifications

Melting Point 199°C to 201°C
Quantity 5 g
Formula Weight 184.64
Percent Purity 99%
Chemical Name or Material 2-Amino-6-chlorobenzothiazole

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.