Learn More
Sigma Laborzentrifugen™ 2-7 Table Centrifuge
Used in biotechnology, pharmacy, medicine, environmental analysis and other industries. Sigma Laborzentrifugen 2-7 Table Centrifuge provides powerful, sustainable and energy-efficient product.
Brand: Sigma Laborzentrifugen™ 10226
2185.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- High quality processing, simple, clear display, large buttons
- Corresponds to most recent safety, emission and environmental criteria
- Fits into laboratory processes integrate
- Comfortable, consumption-optimized and low-noise centrifuges (55dB)
- Space-saving, quiet and easy to use, user-friendly
- Has blue LED run indicator that is visible from distance
- Noise-optimized operation, stainless steel boiler, motorized lid lock
- Spincontrol E offers high comfort with inputs and programs
- Settings can be saved, standstill and start timesare optionally programmable, and can be changed during run (ex: Clotting Time)
- Acoustic signal for pushbutton pressure and current
- Automatic process sequence for gel tubes
- 10 individual programs can be stored and retrieved
- Low temperature rise in continuous operation
- Maintenance-free motor
- 2 Acceleration and braking curves, soft and fast
- Easy changing of rotors
- Unbalance monitoring and disconnection
- Overspeed control
- Visible window in the cover for independent speed measurement
- Manufactured according to EN 61010-2-020, RoHS compliant
- Also used as IVD device according to 98/79/EG in vitro diagnostics
Specifications
12 sec. | |
Microprocessor | |
30 x 15 mL, 4 x 100 mL | |
LED | |
2-7 Table Centrifuge | |
12/12 sec. | |
10 to 99 min. 59 sec. | |
378 mm | |
293 mm | |
100/240 V 50/60 Hz |
EN 61010-2-020, RoHS compliant | |
Rotor not included; sold separately | |
4,000 rpm | |
100 to 4,000 rpm | |
55 dB | |
10 programs | |
2-7 Table Centrifuge | |
11.5 in. | |
50.7 lb. (23 kg) | |
210 w |