Learn More
2,7-Dibromonaphthalene, 99%
Product Code. p-7037376
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
CAS: 58556-75-5 | C10H6Br2 | 285.97 g/mol
It is an intermediate in dyestuffs and pharmaceuticals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is an intermediate in dyestuffs and pharmaceuticals.
Solubility
Soluble in acetonitrile.
Notes
Incompatible with oxidizing agents. Store at room temperature.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 58556-75-5
Molecular Formula C10H6Br2
Molecular Weight (g/mol) 285.97
MDL Number MFCD00238580
InChI Key ODJZWBLNJKNOJK-UHFFFAOYSA-N
Synonym naphthalene, 2,7-dibromo, acmc-209m5e, ksc490o3j, 2,7-dibromonaphthalene hplc
PubChem CID 625355
IUPAC Name 2,7-dibromonaphthalene
SMILES BrC1=CC2=CC(Br)=CC=C2C=C1
Specifications
Melting Point 139°C to 142°C
Quantity 250 mg
UN Number UN3077
Beilstein 1862041
Solubility Information Soluble in acetonitrile.
Formula Weight 285.97
Percent Purity 99%
Chemical Name or Material 2,7-Dibromonaphthalene