Learn More
2,6-Dimethyl-p-benzoquinone, 99%, Thermo Scientific Chemicals
Product Code. 11483754
Click to view available options
Quantity:
1 g
5 g
25 g
2,6-Dimethyl-p-benzoquinone is used as organic intermediates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2,6-Dimethyl-p-benzoquinone is used as organic intermediates.
Solubility
Soluble in chloroform. Insoluble in water.
Notes
Incompatible with strong oxidizing agents and reducing agents.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 527-61-7
Molecular Formula C8H8O2
Molecular Weight (g/mol) 136.15
MDL Number MFCD00001605
InChI Key SENUUPBBLQWHMF-UHFFFAOYSA-N
Synonym 2,6-dimethylbenzoquinone, 2,6-dimethyl-1,4-benzoquinone, 2,6-dimethyl-p-benzoquinone, m-xyloquinone, 2,6-xyloquinone, 2,6-dimethylquinone, 2,5-cyclohexadiene-1,4-dione, 2,6-dimethyl, 3,5-dimethylbenzoquinone, p-benzoquinone, 2,6-dimethyl, 2,6-dimethyl-1,4-quinone
PubChem CID 68241
IUPAC Name 2,6-dimethylcyclohexa-2,5-diene-1,4-dione
SMILES CC1=CC(=O)C=C(C1=O)C
Specifications
Melting Point 69°C to 71°C
Quantity 5 g
Beilstein 2041345
Solubility Information Soluble in chloroform. Insoluble in water.
Formula Weight 136.15
Percent Purity 99%
Chemical Name or Material 2,6-Dimethyl-p-benzoquinone