Learn More
Labconco 100 mL Pear Shaped Flask Rotor - CentriVap Pro
Rotors for use with Labconco™ CentriVap™ Pro Vacuum Concentrators. Aluminum and PTFE coated options available.
Brand: Labconco 7556601
This item is not returnable.
View return policy
Description
- Compatible with CentriVap Pro Vacuum Concentrators
- Aluminum material
- Optional PTFE coated
Specifications
Aluminum | |
PTFE | |
1.8 in. | |
11.4 in. |
Rotor | |
CentriVap Pro | |
4.57 cm | |
28.96 cm |