Learn More
1-(Trimethylsiloxy)cyclohexene, 98%, Thermo Scientific Chemicals Product Code.: 11453127
1-(Trimethylsiloxy)cyclohexene, 98%, Thermo Scientific Chemicals
Product Code. 11453127
250 g
Click to view available options
Quantity:
10 g
50 g
250 g
Product Code. 11453127
Brand: Thermo Scientific Alfa Aesar A16099.30
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
1-(Trimethylsiloxy)cyclohexene is used as intermediate to complex compounds, including natural products. It is also used as a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1-(Trimethylsiloxy)cyclohexene is used as intermediate to complex compounds, including natural products. It is also used as a pharmaceutical intermediate.
Solubility
Not miscible or difficult to mix.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents. It is sensitive to moisture. Store under dry inert gas.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 6651-36-1
Molecular Formula C9H18OSi
Molecular Weight (g/mol) 170.33
MDL Number MFCD00001541
InChI Key SBEMOANGDSSPJY-UHFFFAOYSA-N
Synonym 1-trimethylsilyloxy cyclohexene, cyclohex-1-en-1-yloxy trimethylsilane, 1-trimethylsiloxy cyclohexene, silane, 1-cyclohexen-1-yloxy trimethyl, 1-cyclohexenyloxytrimethylsilane, 1-cyclohexen-1-yloxy trimethylsilane, cyclohexene, 1-trimethylsilyl oxy, cyclohexenyloxy trimethylsilane, 1-trimethylsiloxycyclohexene, cyclohexenyloxytrimethylsilane
PubChem CID 81161
IUPAC Name cyclohexen-1-yloxy(trimethyl)silane
SMILES C[Si](C)(C)OC1=CCCCC1
Specifications
Boiling Point 165°C
Quantity 250 g
UN Number UN1993
Beilstein 1859394
Flash Point 42°C (107°F)
Solubility Information Not miscible or difficult to mix.
Formula Weight 170.33
Percent Purity 98%
Refractive Index 1.447
Sensitivity Moisture sensitive
Density 0.883
Chemical Name or Material 1-(Trimethylsiloxy)cyclohexene
Product Title
1-(Trimethylsiloxy)cyclohexene, 98%, Thermo Scientific Chemicals >