Learn More
1-Indanol, 98%
Product Code. p-7027635
Click to view available options
Quantity:
5 g
25 g
50 g
Unit Size:
25g
50g
5g
CAS: 6351-10-6 | C9H10O | 134.18 g/mol
1-Indanol derivative was found to be an efficient ligand for the palladium-catalyzed asymmetric Heck reaction. It is used as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1-Indanol derivative was found to be an efficient ligand for the palladium-catalyzed asymmetric Heck reaction. It is used as pharmaceutical intermediate.
Solubility
Soluble in water 10 g/L at 20°C, ethanol, benzene.
Notes
Store in a cool, dry place in tightly closed container. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 6351-10-6
Molecular Formula C9H10O
Molecular Weight (g/mol) 134.18
MDL Number MFCD00003797
InChI Key YIAPLDFPUUJILH-UHFFFAOYNA-N
Synonym 1-indanol, indanol, indan-1-ol, 1-hydroxyhydrindene, 1h-inden-1-ol, 2,3-dihydro, 1h-indenol, 2,3-dihydro, 1-hydroxyindan, 1-hydroxyindane, 1-indanole, --indanol
PubChem CID 22819
ChEBI CHEBI:16697
IUPAC Name 2,3-dihydro-1H-inden-1-ol
SMILES C1CC2=CC=CC=C2C1O
Specifications
Melting Point 50°C to 54°C
Boiling Point 128°C (12 mmHg)
Flash Point 145°C (293°F)
Quantity 5 g
Beilstein 2042960
Solubility Information Soluble in water 10g/L at 20°C,ethanol,benzene.
Formula Weight 134.18
Percent Purity 98%
Chemical Name or Material 1-Indanol