Learn More
Sartorius™ Standard Flexboy™ Bioprocessing Bags
Provides a single-use alternative to glass and plastic carboys in bioprocess applications. Sartorius™ Standard Flexboy™ Bioprocessing Bags feature EVA tubing and MPC connections, and are extensively qualified to be safe and robust. Pre-sterilized, ready-to-use bags with volumes between 5 and 50L.
Brand:  Sartorius™ FFB102340
Product Code. 13034373
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
Superior biopharmaceutical fluid handling
  • Ideal for the preparation, storage and transport of biopharmaceuticals, intermediates and final bulk products
  • The broad chemical compatibility assures the safe processing of a wide range of process fluids
  • Single-use alternative to traditional glass, stainless steel and rigid plastic carboys for many applications
  • Ethyl Vinyl Acetate (EVA) tubing for superior flexibility, impact strength and low-temp qualities
  • Three bag ports per bag with easy-to-use and secure MPC male couplings:
    • Bag ports 1 and 2: 3/8 in. x 15/32 in. x 10cm with dust caps and slide clamps
    • Bag port 3: 3/16 in. x ¼ in. x 5cm with a septum
Risk and cost reduction
  • 100% integrity testing of bag and connections to ensure process safety and integrity
  • Full compliance with ISO 11137 for the highest sterility assurance level
  • Single-use system reduces the risk of cross contamination between batches and products
  • Costly and time-consuming CIP and SIP operations are minimized
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Sartorius™ Standard Flexboy™ Bioprocessing Bags > Capacity: 50L