Learn More
1,3-Bis(aminopropyl)tetramethyldisiloxane, 94%
Product Code. p-7047027
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
CAS: 2469-55-8 | C10H28N2OSi2 | 248.52 g/mol
1,3-Bis(aminopropyl)tetramethyldisiloxane is used as a curing agent for epoxy molding compounds, which is utilized for high-reliability semiconductor devices. It is also used to prepare siloxane- urea copymers from 4,4’-diphenylmethane diisocyanate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1,3-Bis(aminopropyl)tetramethyldisiloxane is used as a curing agent for epoxy molding compounds, which is utilized for high-reliability semiconductor devices. It is also used to prepare siloxane- urea copymers from 4,4’-diphenylmethane diisocyanate.
Solubility
Slightly soluble in water.
Notes
Air Sensitive. Incompatible with strong oxidizing agents. Store in a cool place.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 2469-55-8
Molecular Formula C10H28N2OSi2
Molecular Weight (g/mol) 248.52
MDL Number MFCD00039786
InChI Key GPXCORHXFPYJEH-UHFFFAOYSA-N
Synonym 1,3-bis 3-aminopropyl tetramethyldisiloxane, 3,3'-1,1,3,3-tetramethyldisiloxane-1,3-diyl bis propan-1-amine, 1,3-bis 3-aminopropyl-1,1,3,3-tetramethyldisiloxane, tetramethyl-1,3-bis 3-aminopropyl disiloxane, 1-propanamine, 3,3'-1,1,3,3-tetramethyl-1,3-disiloxanediyl bis, oxybis 3-aminopropyl dimethylsilane, bis 3-aminopropyl tetramethyl disiloxane, silane, oxybis 3-aminopropyl dimethyl, bis gamma-aminopropyl tetramethyldisiloxane, bis 3-aminopropyl tetramethyldisiloxane
PubChem CID 17181
IUPAC Name 3-[[3-aminopropyl(dimethyl)silyl]oxy-dimethylsilyl]propan-1-amine
SMILES C[Si](C)(CCCN)O[Si](C)(C)CCCN
Specifications
Boiling Point 132°C to 139°C (11mmHg)
Quantity 25 g
UN Number UN2735
Beilstein 1760876
Flash Point 91°C (195°F)
Solubility Information Slightly soluble in water.
Formula Weight 248.52
Percent Purity 94%
Refractive Index 1.45
Sensitivity Air Sensitive
Density 0.897
Chemical Name or Material 1,3-Bis(aminopropyl)tetramethyldisiloxane