missing translation for 'onlineSavingsMsg'
Learn More

1,2-Diaminocyclohexanetetraacetic acid monohydrate, Honeywell Fluka™

Product Code. 15678370 Shop All Honeywell Fluka Products
Click to view available options
Quantity:
25 g
100 g
6 x 25 g
6 x 100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

for complexometry,≥98.5%

Honeywell´s high-quality titration products cover the complete titration workflow and include:

  • Solutions for volumetric titration (acids, bases, or salts)
  • Buffers available as concentrates or ready-to-use

 

  • Reagents for complexometry:
    • Aminopolycarboxylic acids (EDTA/NTA analogs)
    • Masking agents
  • Indicators for volumetric and complexometric titrations

 


 

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 145819-99-4
Molecular Formula C14H24N2O9
Molecular Weight (g/mol) 364.351
MDL Number MFCD00150952
InChI Key VASZYFIKPKYGNC-UHFFFAOYSA-N
Synonym cdta hydrate, glycine, n,n'-1,2-cyclohexanediylbis n-carboxymethyl-, monohydrate, trans-1,2-cyclohexanediamine-n,n,n',n'-tetraacetic acid monohydrate, acmc-20n4nc, 1,2-diaminocyclohexanetetraacetic acid monohydrate, trans-1,2-cyclohexanediamine-n,n,n',n-tetraacetic acid, 2,2',2,2'-cyclohexane-1,2-diylbis azanetriyl tetraacetic acid hydrate, 2,2',2,2'-cyclohexane-1,2-diyldinitrilo tetraacetic acid-water 1/1, 2-2-bis carboxymethyl amino cyclohexyl-carboxymethyl amino acetic acid hydrate, 1,2-diaminocyclohexanetetraacetic acid monohydrate for complexometry, inverted exclamation marky
PubChem CID 18674933
IUPAC Name 2-[[2-[bis(carboxymethyl)amino]cyclohexyl]-(carboxymethyl)amino]acetic acid;hydrate
SMILES C1CCC(C(C1)N(CC(=O)O)CC(=O)O)N(CC(=O)O)CC(=O)O.O

Specifications

Quantity 6 x 100 g
UN Number NONH for all modes of transport
Formula Weight 364.35
Percent Purity ≥98.5%
Chemical Name or Material 1,2-Diaminocyclohexanetetraacetic acid monohydrate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.