Learn More
VapLock Tubing Adapter, 1.8-2.0 mm OD x 1/4" NPT(M), red PP w/ green ETFE, PP; Cole-Parmer
Brand:  Vaplock AC-151
VapLock Tubing Adapter, 1.8-2.0 mm OD x 1/4" NPT(M), red PP w/ green ETFE, PP; Cole-Parmer
Additional Details : Weight : 0.05000kg
Product Code. 15844404