Hexane (C6H14)
Hexane CHROMASOLV™, for HPLC, ≥97.0% (GC), Honeywell Riedel-de Haën™
CAS: 110-54-3 Molecular Formula: C6H14 Molecular Weight (g/mol): 86.18 MDL Number: MFCD02179311 InChI Key: VLKZOEOYAKHREP-UHFFFAOYSA-N Synonym: hexanes, hexanen, heksan, dipropyl, hexan, hexyl hydride, gettysolve-b, skellysolve b, esani, n-hexane PubChem CID: 8058 ChEBI: CHEBI:29021 IUPAC Name: hexane SMILES: CCCCCC
N-Hexane, Certified AR for Analysis, Fisher Chemical™
C6H14, CAS Number-110-54-3, hexanes, hexyl hydride, hexan, hexanen, gettysolve-b, heksan, dipropyl, esani, n-hexane, skellysolve b, 2.5L, 69 deg.C, CHEBI:29021, Colorless, 86.18g/mol, VLKZOEOYAKHREP-UHFFFAOYSA-N, -95 deg.C, 2179311, 86.178, Plastic Coated Glass bottle, Liquid, 8058, CCCCCC, 2.97
Hexane, 95% n-Hexane, BAKER ANALYZED, J.T. Baker™
CAS: 110-54-3 Molecular Formula: C6H14 Molecular Weight (g/mol): 86.18 MDL Number: MFCD02179311 InChI Key: VLKZOEOYAKHREP-UHFFFAOYSA-N Synonym: hexanes, hexanen, heksan, dipropyl, hexan, hexyl hydride, gettysolve-b, skellysolve b, esani, n-hexane PubChem CID: 8058 ChEBI: CHEBI:29021 IUPAC Name: hexane SMILES: CCCCCC
Hexanes, ≈95% N-Hexane, for HPLC
CAS: 110-54-3 Molecular Formula: C6H14 Molecular Weight (g/mol): 86.18 MDL Number: MFCD02179311 InChI Key: VLKZOEOYAKHREP-UHFFFAOYSA-N Synonym: hexanes, hexanen, heksan, dipropyl, hexan, hexyl hydride, gettysolve-b, skellysolve b, esani, n-hexane PubChem CID: 8058 ChEBI: CHEBI:29021 IUPAC Name: hexane SMILES: CCCCCC
Hexane, Laboratory Reagent, ≥95%, Honeywell™
CAS: 110-54-3 Molecular Formula: C6H14 Molecular Weight (g/mol): 86.18 MDL Number: MFCD02179311 InChI Key: VLKZOEOYAKHREP-UHFFFAOYSA-N Synonym: hexanes, hexanen, heksan, dipropyl, hexan, hexyl hydride, gettysolve-b, skellysolve b, esani, n-hexane PubChem CID: 8058 ChEBI: CHEBI:29021 IUPAC Name: hexane SMILES: CCCCCC
n-Hexane, 99%, Thermo Scientific Chemicals
CAS: 110-54-3 Molecular Formula: C6H14 Molecular Weight (g/mol): 86.18 MDL Number: MFCD02179311 InChI Key: VLKZOEOYAKHREP-UHFFFAOYSA-N Synonym: hexanes, hexanen, heksan, dipropyl, hexan, hexyl hydride, gettysolve-b, skellysolve b, esani, n-hexane PubChem CID: 8058 ChEBI: CHEBI:29021 IUPAC Name: hexane SMILES: CCCCCC
Hexane Fraction, Specified Reagent for General Laboratory Work, SLR, Fisher Chemical™
CAS: 92112-69-1 Molecular Formula: C6H14 Molecular Weight (g/mol): 86.18 MDL Number: MFCD00009520 InChI Key: VLKZOEOYAKHREP-UHFFFAOYSA-N Synonym: hexanes, hexanen, heksan, dipropyl, hexan, hexyl hydride, gettysolve-b, skellysolve b, esani, n-hexane PubChem CID: 8058 ChEBI: CHEBI:29021 IUPAC Name: hexane SMILES: CCCCCC
Isohexane, contains <5% N-Hexane, Extra Pure, SLR
CAS: 73513-42-5 Molecular Formula: C6H14 Molecular Weight (g/mol): 86.18 MDL Number: MFCD00009406 InChI Key: AFABGHUZZDYHJO-UHFFFAOYSA-N Synonym: 2-methyl-pentane, unii-49ib0u6mld, methyl pentane, iso-hexane, 1,1-dimethylbutane, dimethylpropylmethane, 2-methyl pentane, 2-methylpentan, pentane, 2-methyl, isohexane PubChem CID: 7892 IUPAC Name: 2-methylpentane SMILES: CCCC(C)C
n-Hexane, for analysis, contains appr. 95% n-Hexane, Thermo Scientific Chemicals
CAS: 110-54-3 Molecular Formula: C6H14 Molecular Weight (g/mol): 86.18 MDL Number: MFCD02179311 InChI Key: VLKZOEOYAKHREP-UHFFFAOYSA-N Synonym: hexanes, hexanen, heksan, dipropyl, hexan, hexyl hydride, gettysolve-b, skellysolve b, esani, n-hexane PubChem CID: 8058 ChEBI: CHEBI:29021 IUPAC Name: hexane SMILES: CCCCCC
n-Hexane, Environmental Grade, 95+%, Thermo Scientific Chemicals
CAS: 110-54-3 Molecular Formula: C6H14 Molecular Weight (g/mol): 86.18 MDL Number: MFCD02179311 InChI Key: VLKZOEOYAKHREP-UHFFFAOYSA-N Synonym: hexanes, hexanen, heksan, dipropyl, hexan, hexyl hydride, gettysolve-b, skellysolve b, esani, n-hexane PubChem CID: 8058 ChEBI: CHEBI:29021 IUPAC Name: hexane SMILES: CCCCCC
Hexanes, Certified AR for Analysis, 95% N-Hexane Approx, Fisher Chemical™
CAS: 92112-69-1 Molecular Formula: C6H14 Molecular Weight (g/mol): 86.18 MDL Number: MFCD00009520 InChI Key: VLKZOEOYAKHREP-UHFFFAOYSA-N Synonym: hexanes, hexanen, heksan, dipropyl, hexan, hexyl hydride, gettysolve-b, skellysolve b, esani, n-hexane PubChem CID: 8058 ChEBI: CHEBI:29021 IUPAC Name: hexane SMILES: CCCCCC
n-Hexane, 95+%, ACS reagent, Thermo Scientific Chemicals
CAS: 110-54-3 Molecular Formula: C6H14 Molecular Weight (g/mol): 86.18 MDL Number: MFCD02179311 InChI Key: VLKZOEOYAKHREP-UHFFFAOYSA-N IUPAC Name: hexane SMILES: CCCCCC
Hexanes, Optima™ for HPLC and GC, Fisher Chemical™
CAS: 92112-69-1 Molecular Formula: C6H14 Molecular Weight (g/mol): 86.18 MDL Number: MFCD02179311 InChI Key: VLKZOEOYAKHREP-UHFFFAOYSA-N Synonym: hexanes, hexanen, heksan, dipropyl, hexan, hexyl hydride, gettysolve-b, skellysolve b, esani, n-hexane PubChem CID: 8058 ChEBI: CHEBI:29021 IUPAC Name: hexane SMILES: CCCCCC
n-Hexane, 95+%, extra pure, Thermo Scientific Chemicals
CAS: 110-54-3 Molecular Formula: C6H14 Molecular Weight (g/mol): 86.18 MDL Number: MFCD02179311 InChI Key: VLKZOEOYAKHREP-UHFFFAOYSA-N Synonym: hexanes, hexanen, heksan, dipropyl, hexan, hexyl hydride, gettysolve-b, skellysolve b, esani, n-hexane PubChem CID: 8058 ChEBI: CHEBI:29021 IUPAC Name: hexane SMILES: CCCCCC
Hexanes, mixed isomers, ACS, 98.5+%, Thermo Scientific Chemicals
CAS: 92112-69-1 Molecular Formula: C6H14 Molecular Weight (g/mol): 86.18 MDL Number: MFCD00009520 InChI Key: VLKZOEOYAKHREP-UHFFFAOYSA-N Synonym: hexanes, hexanen, heksan, dipropyl, hexan, hexyl hydride, gettysolve-b, skellysolve b, esani, n-hexane PubChem CID: 8058 ChEBI: CHEBI:29021 IUPAC Name: hexane SMILES: CCCCCC
n-Hexane, Spectrophotometric Grade, 95+%, Thermo Scientific Chemicals
CAS: 110-54-3 Molecular Formula: C6H14 Molecular Weight (g/mol): 86.18 MDL Number: MFCD02179311 InChI Key: VLKZOEOYAKHREP-UHFFFAOYSA-N Synonym: hexanes, hexanen, heksan, dipropyl, hexan, hexyl hydride, gettysolve-b, skellysolve b, esani, n-hexane PubChem CID: 8058 ChEBI: CHEBI:29021 IUPAC Name: hexane SMILES: CCCCCC
Hexanes, mixed isomers, Environmental Grade, Thermo Scientific Chemicals
CAS: 92112-69-1 Molecular Formula: C6H14 Molecular Weight (g/mol): 86.18 MDL Number: MFCD00009520 InChI Key: VLKZOEOYAKHREP-UHFFFAOYSA-N Synonym: hexanes, hexanen, heksan, dipropyl, hexan, hexyl hydride, gettysolve-b, skellysolve b, esani, n-hexane PubChem CID: 8058 ChEBI: CHEBI:29021 IUPAC Name: hexane SMILES: CCCCCC
n-Hexane, 99+%, for analysis, Thermo Scientific Chemicals
CAS: 110-54-3 Molecular Formula: C6H14 Molecular Weight (g/mol): 86.18 MDL Number: MFCD02179311 InChI Key: VLKZOEOYAKHREP-UHFFFAOYSA-N Synonym: hexanes, hexanen, heksan, dipropyl, hexan, hexyl hydride, gettysolve-b, skellysolve b, esani, n-hexane PubChem CID: 8058 ChEBI: CHEBI:29021 IUPAC Name: hexane SMILES: CCCCCC
Hexanes (Certified ACS), Fisher Chemical™
CAS: 92112-69-1 Molecular Formula: C6H14 Molecular Weight (g/mol): 86.18 MDL Number: MFCD02179311 InChI Key: VLKZOEOYAKHREP-UHFFFAOYSA-N Synonym: hexanes, hexanen, heksan, dipropyl, hexan, hexyl hydride, gettysolve-b, skellysolve b, esani, n-hexane PubChem CID: 8058 ChEBI: CHEBI:29021 IUPAC Name: hexane SMILES: CCCCCC
Hexanes, mixed isomers, (60+% n-hexane), Thermo Scientific Chemicals
CAS: 92112-69-1 Molecular Formula: C6H14 Molecular Weight (g/mol): 86.18 MDL Number: MFCD00009520 InChI Key: VLKZOEOYAKHREP-UHFFFAOYSA-N Synonym: hexanes, hexanen, heksan, dipropyl, hexan, hexyl hydride, gettysolve-b, skellysolve b, esani, n-hexane PubChem CID: 8058 ChEBI: CHEBI:29021 IUPAC Name: hexane SMILES: CCCCCC
n-Hexane, Spectrophotometric Grade, 95+%, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific Chemicals
CAS: 110-54-3 Molecular Formula: C6H14 Molecular Weight (g/mol): 86.18 MDL Number: MFCD02179311 InChI Key: VLKZOEOYAKHREP-UHFFFAOYSA-N Synonym: hexanes, hexanen, heksan, dipropyl, hexan, hexyl hydride, gettysolve-b, skellysolve b, esani, n-hexane PubChem CID: 8058 ChEBI: CHEBI:29021 IUPAC Name: hexane SMILES: CCCCCC
n-hexane, 95+%, for HPLC, Thermo Scientific Chemicals
CAS: 110-54-3 Molecular Formula: C6H14 Molecular Weight (g/mol): 86.18 MDL Number: MFCD02179311 InChI Key: VLKZOEOYAKHREP-UHFFFAOYSA-N Synonym: hexanes, hexanen, heksan, dipropyl, hexan, hexyl hydride, gettysolve-b, skellysolve b, esani, n-hexane PubChem CID: 8058 ChEBI: CHEBI:29021 IUPAC Name: hexane SMILES: CCCCCC
n-Hexane, 97%, Extra Dry over Molecular Sieve, AcroSeal™, Thermo Scientific Chemicals
CAS: 110-54-3 Molecular Formula: C6H14 Molecular Weight (g/mol): 86.18 MDL Number: MFCD02179311 InChI Key: VLKZOEOYAKHREP-UHFFFAOYSA-N Synonym: hexanes, hexanen, heksan, dipropyl, hexan, hexyl hydride, gettysolve-b, skellysolve b, esani, n-hexane PubChem CID: 8058 ChEBI: CHEBI:29021 IUPAC Name: hexane SMILES: CCCCCC
The term “hexanes” (CAS 110-54-3) refers to a hydrocarbon distillation fraction that contains a mixture of linear and branched hexane isomers. Hexanes typically contain a high proportion of n-hexane; some 2-methylpentane, 3-methylpentane, 2,3-dimethylbutane, and 2,2,-dimethylbutane; and smaller amounts of n-pentane, cyclohexane, n-heptane, and methylcyclopentane.
Hexanes are a clear, colorless, highly flammable liquid with a gasoline-like odor (although odorless when pure). Insoluble in water, it is soluble in ether, alcohol, and acetone.
Hexanes are used in the laboratory to extract oil and grease contaminants from water and soil for analysis, as a non-polar chromatography solvent, and to create organolithiums and other chemical reactions that involve very strong bases.
In industrial settings, hexanes are used as solvents to extract edible oils from seeds and vegetables, as cleaning agents (in the textile, furniture, and printing industries), and as glues in roofing, shoemaking, and leather product manufacturing.
Inhalation exposure can affect the nervous system and cause dizziness, nausea, and headache; prolonged contact can cause eye and skin irritation. Hexanes are flammable — keep away from heat sources and store in a closed environment with proper ventilation.